The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002917.1 | NP_001002917.1 | 1 - 1 | 629 | 101 |
GenBank mRNA | AF238489.1 | - | 1 - 1 | 629 | 101 |
BC136950.1 | - | 1 - 1 | 700 | 101 | |
BC136954.1 | - | 1 - 1 | 700 | 101 |