The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001969.4 | NP_001960.2 | 8 - 9 | 1419 | 139 |
NM_183004.4 | NP_892116.2 | 7 - 8 | 1276 | 139 | |
GenBank mRNA | AK026933.1 | - | 8 - 9 | 1211 | 139 |
AL080102.1 | - | 8 - 9 | 1219 | 139 | |
BC007728.2 | - | 7 - 8 | 1160 | 139 | |
BC032866.2 | - | 7 - 8 | 1227 | 139 | |
BX537367.1 | - | 6 - 7 | 1655 | 139 | |
CR456712.1 | - | 6 - 7 | 743 | 139 | |
U49436.1 | - | 7 - 8 | 1062 | 139 |