The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270965.1 | NP_001257894.1 | 10 - 11 | 2199 | 78 |
NM_001270966.1 | NP_001257895.1 | 10 - 11 | 1720 | 78 | |
NM_002779.4 | NP_002770.3 | 9 - 10 | 2523 | 78 | |
XM_011539968.1 | XP_011538270.1 | 10 - 11 | 2205 | 78 | |
XM_011539969.2 | XP_011538271.1 | 10 - 11 | 2222 | 78 | |
XM_017016433.1 | XP_016871922.1 | 10 - 11 | 2216 | 78 | |
XM_017016434.1 | XP_016871923.1 | 9 - 10 | 2143 | 78 | |
GenBank mRNA | AB095931.1 | - | 9 - 10 | 2523 | 78 |
BC039259.1 | - | 10 - 11 | 1707 | 78 | |
BC142643.1 | - | 10 - 11 | 2199 | 78 | |
BC142689.1 | - | 10 - 11 | 2199 | 78 |