Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSD
Entrez Gene ID
5662
Gene Name
pleckstrin and Sec7 domain containing
Gene Aliases
EFA6, EFA6A, PSD1, TYL
Chromosome Location
Chr.10: 102402617 - 102420164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270965.1 NP_001257894.1 14 - 15 2602 76
NM_001270966.1 NP_001257895.1 14 - 15 2123 76
NM_002779.4 NP_002770.3 13 - 14 2926 76
NR_073110.1 4 - 5 698 76
XM_011539968.1 XP_011538270.1 14 - 15 2608 76
XM_011539969.2 XP_011538271.1 14 - 15 2625 76
XM_017016433.1 XP_016871922.1 14 - 15 2619 76
XM_017016434.1 XP_016871923.1 13 - 14 2546 76
GenBank mRNA AB095931.1 - 13 - 14 2926 76
BC039259.1 - 14 - 15 2110 76
BC142643.1 - 14 - 15 2602 76
BC142689.1 - 14 - 15 2602 76
BM920983.1 - - 429 76

View Full Product Details