The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077494.3 | NP_001070962.1 | 21 - 22 | 2738 | 94 |
NM_001261403.2 | NP_001248332.1 | 20 - 21 | 2833 | 94 | |
NM_001288724.1 | NP_001275653.1 | 21 - 22 | 3032 | 94 | |
NM_001322934.1 | NP_001309863.1 | 21 - 22 | 2742 | 94 | |
NM_001322935.1 | NP_001309864.1 | 20 - 21 | 2906 | 94 | |
NM_002502.5 | NP_002493.3 | 21 - 22 | 2742 | 94 | |
XM_011539830.2 | XP_011538132.1 | 16 - 17 | 2096 | 94 | |
XM_011539831.2 | XP_011538133.1 | 16 - 17 | 2093 | 94 | |
XM_017016278.1 | XP_016871767.1 | 21 - 22 | 3027 | 94 | |
GenBank mRNA | AK098415.1 | - | 11 - 12 | 1615 | 94 |
AK292654.1 | - | 20 - 21 | 2686 | 94 | |
BC002844.2 | - | 21 - 22 | 2569 | 94 | |
BT009769.1 | - | 20 - 21 | 2469 | 94 | |
S76638.1 | - | 21 - 22 | 2719 | 94 | |
X61498.1 | - | 21 - 22 | 2629 | 94 |