The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077494.3 | NP_001070962.1 | 5 - 6 | 509 | 59 |
NM_001261403.2 | NP_001248332.1 | 4 - 5 | 604 | 59 | |
NM_001288724.1 | NP_001275653.1 | 5 - 6 | 803 | 59 | |
NM_001322934.1 | NP_001309863.1 | 5 - 6 | 513 | 59 | |
NM_001322935.1 | NP_001309864.1 | 5 - 6 | 803 | 59 | |
NM_002502.5 | NP_002493.3 | 5 - 6 | 513 | 59 | |
XM_017016278.1 | XP_016871767.1 | 5 - 6 | 798 | 59 | |
GenBank mRNA | AK292654.1 | - | 4 - 5 | 457 | 59 |
BC002844.2 | - | 5 - 6 | 340 | 59 | |
BT009769.1 | - | 4 - 5 | 240 | 59 | |
BY799958.2 | - | 5 - 6 | 441 | 59 | |
DA408461.1 | - | 5 - 6 | 440 | 59 | |
DA711063.1 | - | 5 - 6 | 507 | 59 | |
S76638.1 | - | 5 - 6 | 490 | 59 | |
U09609.2 | - | 4 - 5 | 240 | 59 | |
X61498.1 | - | 5 - 6 | 403 | 59 | |
X61499.1 | - | 5 - 6 | 403 | 59 |