The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134444.4 | NP_604393.2 | 4 - 5 | 2440 | 88 |
XM_017026344.1 | XP_016881833.1 | 3 - 4 | 2083 | 88 | |
XM_017026345.1 | XP_016881834.1 | 3 - 4 | 2083 | 88 | |
GenBank mRNA | AF442488.1 | - | 3 - 4 | 2075 | 88 |
AF479747.1 | - | 3 - 4 | 2087 | 88 | |
AF482706.1 | - | 3 - 4 | 2085 | 88 | |
AK056688.1 | - | 2 - 3 | 1967 | 88 | |
AY072792.1 | - | 3 - 4 | 2078 | 88 | |
BC016443.1 | - | 2 - 3 | 715 | 88 | |
BC050326.1 | - | 4 - 5 | 2114 | 88 |