Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP8
Entrez Gene ID
155038
Gene Name
GTPase, IMAP family member 8
Gene Aliases
IAN-9, IAN6, IAN9, IANT
Chromosome Location
Chr.7: 150450630 - 150479395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175571.3 NP_783161.1 4 - 5 1884 75
XM_005249950.4 XP_005250007.1 4 - 5 1564 75
GenBank mRNA AJ633686.1 - 3 - 4 1310 75
AK098320.1 - 3 - 4 1070 75
AK314426.1 - 4 - 5 1720 75
AL834361.1 - 4 - 5 1478 75
BC107037.2 - 4 - 5 1471 75

View Full Product Details