The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007269.2 | NP_009200.2 | 15 - 16 | 1434 | 68 |
GenBank mRNA | AF032922.1 | - | 15 - 16 | 1416 | 68 |
AK223170.1 | - | 15 - 16 | 1413 | 68 | |
AK290134.1 | - | 15 - 16 | 1418 | 68 | |
AK291322.1 | - | 15 - 16 | 1414 | 68 | |
AK312910.1 | - | 15 - 16 | 1411 | 68 | |
BC038099.2 | - | 15 - 16 | 1373 | 68 | |
BC047764.1 | - | 15 - 16 | 1409 | 68 | |
D63506.1 | - | 15 - 16 | 1410 | 68 |