Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASP7
Entrez Gene ID
840
Gene Name
caspase 7
Gene Aliases
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3
Chromosome Location
Chr.10: 113679162 - 113730909 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001227.4 NP_001218.1 5 - 6 864 114
NM_001267056.1 NP_001253985.1 5 - 6 742 114
NM_001267057.1 NP_001253986.1 5 - 6 895 114
NM_001267058.1 NP_001253987.1 4 - 5 546 114
NM_001320911.1 NP_001307840.1 5 - 6 773 114
NM_033338.5 NP_203124.1 6 - 7 971 114
NM_033339.4 NP_203125.1 6 - 7 938 114
XM_006718017.3 XP_006718080.1 6 - 7 675 114
XM_011540260.1 XP_011538562.1 4 - 5 894 114
XM_017016763.1 XP_016872252.1 5 - 6 922 114
XM_017016764.1 XP_016872253.1 5 - 6 689 114
GenBank mRNA AB451281.1 - 5 - 6 653 114
AK223489.1 - 5 - 6 635 114
AK298964.1 - 4 - 5 507 114
BC015799.1 - 5 - 6 632 114
BT006683.1 - 4 - 5 554 114
BX647973.1 - 5 - 6 673 114
DC332144.1 - 4 - 5 547 114
U37448.1 - 5 - 6 597 114
U40281.1 - 5 - 6 601 114
U67206.1 - 6 - 7 940 114
U67320.1 - 5 - 6 593 114

View Full Product Details