The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324504.1 | NP_001311433.1 | 7 - 8 | 654 | 64 |
NM_001324505.1 | NP_001311434.1 | 5 - 6 | 622 | 64 | |
NM_001324506.1 | NP_001311435.1 | 4 - 5 | 528 | 64 | |
NM_020382.4 | NP_065115.3 | 6 - 7 | 744 | 64 | |
XM_006719394.3 | XP_006719457.1 | 5 - 6 | 574 | 64 | |
XM_011538333.2 | XP_011536635.1 | 5 - 6 | 960 | 64 | |
GenBank mRNA | AK292645.1 | - | 6 - 7 | 712 | 64 |
AY064546.1 | - | 1 - 1 | 796 | 64 | |
BC050346.1 | - | 6 - 7 | 1087 | 64 | |
HQ259683.1 | - | 6 - 7 | 571 | 64 |