The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002901.2 | NP_002892.1 | 4 - 5 | 961 | 89 |
GenBank mRNA | AK094360.1 | - | 3 - 4 | 1051 | 89 |
AK129791.1 | - | 4 - 5 | 988 | 89 | |
BC010120.2 | - | 4 - 5 | 830 | 89 | |
D42073.1 | - | 4 - 5 | 747 | 89 | |
FJ224346.1 | - | 4 - 5 | 747 | 89 |