The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017925.6 | NP_060395.5 | 8 - 9 | 1258 | 68 |
NR_073201.3 | 8 - 9 | 1258 | 68 | ||
XM_006716804.3 | XP_006716867.1 | 8 - 9 | 1226 | 68 | |
XM_006716805.3 | XP_006716868.1 | 8 - 9 | 1226 | 68 | |
XM_011517959.2 | XP_011516261.1 | 8 - 9 | 1226 | 68 | |
XM_017014917.1 | XP_016870406.1 | 8 - 9 | 1226 | 68 | |
XM_017014918.1 | XP_016870407.1 | 9 - 10 | 1426 | 68 | |
XM_017014919.1 | XP_016870408.1 | 9 - 10 | 1393 | 68 | |
XM_017014920.1 | XP_016870409.1 | 9 - 10 | 1426 | 68 | |
GenBank mRNA | AK125842.1 | - | 2 - 3 | 120 | 68 |
BC144484.1 | - | 4 - 5 | 490 | 68 | |
BC144485.1 | - | 4 - 5 | 490 | 68 |