The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004295.3 | NP_004286.2 | 3 - 4 | 405 | 92 |
XM_011525504.2 | XP_011523806.1 | 3 - 4 | 428 | 92 | |
XM_011525505.2 | XP_011523807.1 | 3 - 4 | 2223 | 92 | |
XM_011525506.2 | XP_011523808.1 | 3 - 4 | 2200 | 92 | |
XM_011525507.2 | XP_011523809.1 | 3 - 4 | 774 | 92 | |
XM_011525508.2 | XP_011523810.1 | 3 - 4 | 491 | 92 | |
GenBank mRNA | AF082185.1 | - | 3 - 4 | 297 | 92 |
AK300804.1 | - | 3 - 4 | 370 | 92 | |
AY937224.1 | - | 3 - 4 | 325 | 92 | |
BC001769.1 | - | 3 - 4 | 389 | 92 | |
CR990949.1 | - | 3 - 4 | 426 | 92 | |
X80200.1 | - | 3 - 4 | 382 | 92 |