The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018169.3 | NP_060639.3 | 2 - 3 | 173 | 120 |
XM_006719099.2 | XP_006719162.1 | 2 - 3 | 792 | 120 | |
XM_011520722.1 | XP_011519024.1 | 2 - 3 | 333 | 120 | |
GenBank mRNA | AK092399.1 | - | 2 - 3 | 143 | 120 |
BC140819.1 | - | 2 - 3 | 124 | 120 | |
BP872820.1 | - | 2 - 3 | 173 | 120 |