Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHX6
Entrez Gene ID
26468
Gene Name
LIM homeobox 6
Gene Aliases
LHX6.1
Chromosome Location
Chr.9: 122185626 - 122228893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242333.1 NP_001229262.1 3 - 4 377 57
NM_001242334.1 NP_001229263.1 2 - 3 361 57
NM_014368.4 NP_055183.2 3 - 4 411 57
NM_199160.3 NP_954629.2 3 - 4 411 57
XM_005251916.3 XP_005251973.1 3 - 4 492 57
XM_011518521.2 XP_011516823.1 3 - 4 492 57
XM_011518522.2 XP_011516824.1 3 - 4 760 57
GenBank mRNA AB031041.1 - 2 - 3 258 57
AB031042.1 - 2 - 3 258 57
AK289827.1 - 2 - 3 361 57
AK297175.1 - 3 - 4 377 57
AK299709.1 - 3 - 4 445 57
AK313808.1 - 2 - 3 301 57
AL136570.1 - 3 - 4 304 57
BC103936.1 - 3 - 4 270 57
BC103937.1 - 3 - 4 270 57
BC103938.1 - 3 - 4 270 57

View Full Product Details