Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHX6
Entrez Gene ID
26468
Gene Name
LIM homeobox 6
Gene Aliases
LHX6.1
Chromosome Location
Chr.9: 122185626 - 122228893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242334.1 NP_001229263.1 7 - 8 1067 90
NM_001242335.1 NP_001229264.1 5 - 6 1276 90
NM_014368.4 NP_055183.2 8 - 9 1117 90
GenBank mRNA AB031041.1 - 7 - 8 964 90
AK126982.1 - 5 - 6 1276 90
AK289827.1 - 7 - 8 1067 90
AK299709.1 - 8 - 9 1151 90
AK313808.1 - 7 - 8 1007 90
AL136570.1 - 8 - 9 1010 90
BC103937.1 - 8 - 9 976 90
BC103938.1 - 8 - 9 973 90

View Full Product Details