The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144636.2 | NP_653237.1 | 2 - 3 | 382 | 90 |
GenBank mRNA | AK056271.1 | - | 2 - 3 | 333 | 90 |