The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105518.1 | NP_001098988.1 | 13 - 14 | 2092 | 174 |
NM_017673.6 | NP_060143.4 | 13 - 14 | 2143 | 174 | |
XM_005245271.1 | XP_005245328.1 | 13 - 14 | 2150 | 174 | |
XM_005245273.1 | XP_005245330.1 | 13 - 14 | 2051 | 174 | |
XM_011509657.1 | XP_011507959.1 | 13 - 14 | 2130 | 174 | |
XM_017001527.1 | XP_016857016.1 | 13 - 14 | 2030 | 174 | |
XM_017001528.1 | XP_016857017.1 | 13 - 14 | 2150 | 174 | |
XM_017001529.1 | XP_016857018.1 | 14 - 15 | 2355 | 174 | |
XM_017001530.1 | XP_016857019.1 | 13 - 14 | 2150 | 174 | |
GenBank mRNA | AF288392.1 | - | 12 - 13 | 1989 | 174 |
AK000128.1 | - | 1 - 2 | 142 | 174 | |
AK093263.1 | - | 7 - 8 | 855 | 174 | |
AK127980.1 | - | 13 - 14 | 2030 | 174 | |
AK225238.1 | - | 1 - 2 | 142 | 174 | |
AK291443.1 | - | 13 - 14 | 2009 | 174 | |
AK315802.1 | - | 13 - 14 | 2110 | 174 | |
BC030781.2 | - | 13 - 14 | 2145 | 174 |