Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRCAM
Entrez Gene ID
4897
Gene Name
neuronal cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.7: 108147626 - 108456442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037132.2 NP_001032209.1 15 - 16 2005 93
NM_001193582.1 NP_001180511.1 18 - 19 2385 93
NM_001193583.1 NP_001180512.1 17 - 18 2328 93
NM_001193584.1 NP_001180513.1 17 - 18 2328 93
XM_005250373.2 XP_005250430.1 18 - 19 2385 93
XM_005250383.2 XP_005250440.1 18 - 19 2385 93
XM_005250385.2 XP_005250442.1 18 - 19 2385 93
XM_006716003.1 XP_006716066.1 18 - 19 2385 93
XM_006716007.1 XP_006716070.1 18 - 19 2385 93
XM_006716012.1 XP_006716075.1 18 - 19 2385 93
XM_006716014.1 XP_006716077.1 18 - 19 2385 93
XM_011516253.1 XP_011514555.1 18 - 19 2385 93
XM_011516255.1 XP_011514557.1 17 - 18 2140 93
XM_011516256.1 XP_011514558.1 18 - 19 2385 93
XM_011516257.1 XP_011514559.1 17 - 18 2367 93
XM_011516258.1 XP_011514560.1 16 - 17 2122 93
XM_011516262.1 XP_011514564.1 17 - 18 2328 93
XM_011516265.2 XP_011514567.1 18 - 19 2385 93
XM_011516266.2 XP_011514568.1 18 - 19 2385 93
XM_011516267.1 XP_011514569.1 18 - 19 2385 93
XM_011516268.2 XP_011514570.1 16 - 17 2200 93
XM_011516269.1 XP_011514571.1 18 - 19 2385 93
XM_011516270.1 XP_011514572.1 18 - 19 2385 93
XM_017012237.1 XP_016867726.1 18 - 19 2385 93
XM_017012238.1 XP_016867727.1 17 - 18 2328 93
XM_017012239.1 XP_016867728.1 16 - 17 2310 93
XM_017012240.1 XP_016867729.1 17 - 18 2328 93
XM_017012241.1 XP_016867730.1 16 - 17 2052 93
XM_017012242.1 XP_016867731.1 16 - 17 2052 93
XM_017012243.1 XP_016867732.1 18 - 19 2385 93
XM_017012244.1 XP_016867733.1 18 - 19 2385 93
XM_017012245.1 XP_016867734.1 18 - 19 2385 93
XM_017012247.1 XP_016867736.1 17 - 18 2328 93
XM_017012249.1 XP_016867738.1 17 - 18 2328 93
XM_017012250.1 XP_016867739.1 16 - 17 2052 93
XM_017012253.1 XP_016867742.1 15 - 16 2054 93
XM_017012255.1 XP_016867744.1 15 - 16 2095 93
XM_017012256.1 XP_016867745.1 15 - 16 2054 93
XM_017012257.1 XP_016867746.1 16 - 17 2083 93
GenBank mRNA AB002341.1 - 17 - 18 2256 93
AJ001054.1 - 3 - 4 405 93
AJ001057.1 - 15 - 16 1884 93
AK299870.1 - 18 - 19 2373 93
BC114570.1 - 17 - 18 2252 93
BC115736.1 - 17 - 18 2252 93

View Full Product Details