Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRCAM
Entrez Gene ID
4897
Gene Name
neuronal cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.7: 108147626 - 108456442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037132.2 NP_001032209.1 17 - 18 2139 70
NM_001193582.1 NP_001180511.1 20 - 21 2519 70
NM_001193583.1 NP_001180512.1 19 - 20 2462 70
NM_001193584.1 NP_001180513.1 19 - 20 2462 70
NM_005010.4 NP_005001.3 18 - 19 2471 70
XM_005250373.2 XP_005250430.1 20 - 21 2519 70
XM_005250383.2 XP_005250440.1 20 - 21 2519 70
XM_005250385.2 XP_005250442.1 20 - 21 2519 70
XM_006716003.1 XP_006716066.1 20 - 21 2519 70
XM_006716007.1 XP_006716070.1 20 - 21 2519 70
XM_006716012.1 XP_006716075.1 20 - 21 2519 70
XM_006716014.1 XP_006716077.1 20 - 21 2519 70
XM_011516253.1 XP_011514555.1 20 - 21 2519 70
XM_011516255.1 XP_011514557.1 19 - 20 2274 70
XM_011516256.1 XP_011514558.1 20 - 21 2519 70
XM_011516257.1 XP_011514559.1 19 - 20 2501 70
XM_011516258.1 XP_011514560.1 18 - 19 2256 70
XM_011516259.1 XP_011514561.1 19 - 20 2489 70
XM_011516261.1 XP_011514563.1 18 - 19 2471 70
XM_011516262.1 XP_011514564.1 19 - 20 2462 70
XM_011516265.2 XP_011514567.1 20 - 21 2519 70
XM_011516266.2 XP_011514568.1 20 - 21 2519 70
XM_011516267.1 XP_011514569.1 20 - 21 2519 70
XM_011516268.2 XP_011514570.1 18 - 19 2334 70
XM_011516269.1 XP_011514571.1 20 - 21 2519 70
XM_011516270.1 XP_011514572.1 20 - 21 2519 70
XM_011516271.1 XP_011514573.1 18 - 19 2471 70
XM_017012236.1 XP_016867725.1 19 - 20 2489 70
XM_017012237.1 XP_016867726.1 20 - 21 2519 70
XM_017012238.1 XP_016867727.1 19 - 20 2462 70
XM_017012239.1 XP_016867728.1 18 - 19 2444 70
XM_017012240.1 XP_016867729.1 19 - 20 2462 70
XM_017012241.1 XP_016867730.1 18 - 19 2186 70
XM_017012242.1 XP_016867731.1 18 - 19 2186 70
XM_017012243.1 XP_016867732.1 20 - 21 2519 70
XM_017012244.1 XP_016867733.1 20 - 21 2519 70
XM_017012245.1 XP_016867734.1 20 - 21 2519 70
XM_017012246.1 XP_016867735.1 19 - 20 2489 70
XM_017012247.1 XP_016867736.1 19 - 20 2462 70
XM_017012248.1 XP_016867737.1 19 - 20 2489 70
XM_017012249.1 XP_016867738.1 19 - 20 2462 70
XM_017012250.1 XP_016867739.1 18 - 19 2186 70
XM_017012251.1 XP_016867740.1 19 - 20 2489 70
XM_017012252.1 XP_016867741.1 19 - 20 2489 70
XM_017012253.1 XP_016867742.1 17 - 18 2188 70
XM_017012254.1 XP_016867743.1 19 - 20 2489 70
XM_017012255.1 XP_016867744.1 17 - 18 2229 70
XM_017012256.1 XP_016867745.1 17 - 18 2188 70
XM_017012257.1 XP_016867746.1 18 - 19 2217 70
XM_017012258.1 XP_016867747.1 18 - 19 2213 70
XM_017012259.1 XP_016867748.1 16 - 17 2158 70
GenBank mRNA AB002341.1 - 19 - 20 2390 70
AK299870.1 - 20 - 21 2507 70
BC098401.1 - 18 - 19 2459 70
BC114570.1 - 19 - 20 2386 70
BC115736.1 - 19 - 20 2386 70
BX538010.1 - 18 - 19 2416 70

View Full Product Details