Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRCAM
Entrez Gene ID
4897
Gene Name
neuronal cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.7: 108147626 - 108456442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193582.1 NP_001180511.1 2 - 3 307 145
NM_001193583.1 NP_001180512.1 2 - 3 307 145
NM_001193584.1 NP_001180513.1 2 - 3 307 145
NM_005010.4 NP_005001.3 2 - 3 307 145
XM_005250373.2 XP_005250430.1 2 - 3 307 145
XM_005250383.2 XP_005250440.1 2 - 3 307 145
XM_005250385.2 XP_005250442.1 2 - 3 307 145
XM_006716003.1 XP_006716066.1 2 - 3 307 145
XM_006716007.1 XP_006716070.1 2 - 3 307 145
XM_006716012.1 XP_006716075.1 2 - 3 307 145
XM_006716014.1 XP_006716077.1 2 - 3 307 145
XM_011516253.1 XP_011514555.1 2 - 3 307 145
XM_011516256.1 XP_011514558.1 2 - 3 307 145
XM_011516257.1 XP_011514559.1 2 - 3 307 145
XM_011516259.1 XP_011514561.1 2 - 3 307 145
XM_011516261.1 XP_011514563.1 2 - 3 307 145
XM_011516262.1 XP_011514564.1 2 - 3 307 145
XM_011516265.2 XP_011514567.1 2 - 3 307 145
XM_011516266.2 XP_011514568.1 2 - 3 307 145
XM_011516267.1 XP_011514569.1 2 - 3 307 145
XM_011516269.1 XP_011514571.1 2 - 3 307 145
XM_011516270.1 XP_011514572.1 2 - 3 307 145
XM_011516271.1 XP_011514573.1 2 - 3 307 145
XM_017012236.1 XP_016867725.1 2 - 3 307 145
XM_017012237.1 XP_016867726.1 2 - 3 307 145
XM_017012238.1 XP_016867727.1 2 - 3 307 145
XM_017012239.1 XP_016867728.1 2 - 3 307 145
XM_017012240.1 XP_016867729.1 2 - 3 307 145
XM_017012243.1 XP_016867732.1 2 - 3 307 145
XM_017012244.1 XP_016867733.1 2 - 3 307 145
XM_017012245.1 XP_016867734.1 2 - 3 307 145
XM_017012246.1 XP_016867735.1 2 - 3 307 145
XM_017012247.1 XP_016867736.1 2 - 3 307 145
XM_017012248.1 XP_016867737.1 2 - 3 307 145
XM_017012249.1 XP_016867738.1 2 - 3 307 145
XM_017012251.1 XP_016867740.1 2 - 3 307 145
XM_017012252.1 XP_016867741.1 2 - 3 307 145
XM_017012254.1 XP_016867743.1 2 - 3 307 145
GenBank mRNA AK299870.1 - 2 - 3 295 145
BC098401.1 - 2 - 3 295 145
BC114570.1 - 2 - 3 231 145
BC115736.1 - 2 - 3 231 145
BX538010.1 - 2 - 3 252 145
BX954399.1 - 2 - 3 292 145
DA292156.1 - 2 - 3 307 145

View Full Product Details