Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRCAM
Entrez Gene ID
4897
Gene Name
neuronal cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.7: 108147626 - 108456442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037132.2 NP_001032209.1 28 - 29 3702 108
NM_001193582.1 NP_001180511.1 29 - 30 3803 108
NM_001193583.1 NP_001180512.1 28 - 29 3746 108
NM_001193584.1 NP_001180513.1 27 - 28 3710 108
NM_005010.4 NP_005001.3 26 - 27 3719 108
XM_005250373.2 XP_005250430.1 31 - 32 4082 108
XM_005250383.2 XP_005250440.1 29 - 30 3812 108
XM_005250385.2 XP_005250442.1 28 - 29 3767 108
XM_006716003.1 XP_006716066.1 31 - 32 4082 108
XM_006716007.1 XP_006716070.1 30 - 31 4046 108
XM_006716012.1 XP_006716075.1 29 - 30 3803 108
XM_006716014.1 XP_006716077.1 28 - 29 3767 108
XM_011516253.1 XP_011514555.1 31 - 32 4091 108
XM_011516255.1 XP_011514557.1 30 - 31 3846 108
XM_011516256.1 XP_011514558.1 31 - 32 4091 108
XM_011516257.1 XP_011514559.1 30 - 31 4073 108
XM_011516258.1 XP_011514560.1 29 - 30 3828 108
XM_011516259.1 XP_011514561.1 30 - 31 4061 108
XM_011516261.1 XP_011514563.1 29 - 30 4043 108
XM_011516262.1 XP_011514564.1 30 - 31 4034 108
XM_011516265.2 XP_011514567.1 31 - 32 4091 108
XM_011516266.2 XP_011514568.1 31 - 32 4091 108
XM_011516267.1 XP_011514569.1 29 - 30 3812 108
XM_011516268.2 XP_011514570.1 29 - 30 3906 108
XM_011516269.1 XP_011514571.1 28 - 29 3776 108
XM_011516270.1 XP_011514572.1 28 - 29 3776 108
XM_011516271.1 XP_011514573.1 26 - 27 3728 108
XM_017012236.1 XP_016867725.1 30 - 31 4052 108
XM_017012237.1 XP_016867726.1 30 - 31 4046 108
XM_017012238.1 XP_016867727.1 30 - 31 4034 108
XM_017012239.1 XP_016867728.1 29 - 30 4016 108
XM_017012240.1 XP_016867729.1 30 - 31 4025 108
XM_017012241.1 XP_016867730.1 29 - 30 3749 108
XM_017012242.1 XP_016867731.1 28 - 29 3713 108
XM_017012243.1 XP_016867732.1 30 - 31 3929 108
XM_017012244.1 XP_016867733.1 30 - 31 3938 108
XM_017012245.1 XP_016867734.1 30 - 31 3929 108
XM_017012246.1 XP_016867735.1 28 - 29 3773 108
XM_017012247.1 XP_016867736.1 28 - 29 3746 108
XM_017012248.1 XP_016867737.1 27 - 28 3746 108
XM_017012249.1 XP_016867738.1 28 - 29 3755 108
XM_017012250.1 XP_016867739.1 27 - 28 3479 108
XM_017012251.1 XP_016867740.1 27 - 28 3737 108
XM_017012252.1 XP_016867741.1 27 - 28 3746 108
XM_017012253.1 XP_016867742.1 25 - 26 3436 108
XM_017012254.1 XP_016867743.1 27 - 28 3737 108
XM_017012255.1 XP_016867744.1 28 - 29 3792 108
XM_017012256.1 XP_016867745.1 25 - 26 3436 108
XM_017012257.1 XP_016867746.1 27 - 28 3501 108
XM_017012258.1 XP_016867747.1 26 - 27 3461 108
XM_017012259.1 XP_016867748.1 24 - 25 3406 108
GenBank mRNA AB002341.1 - 27 - 28 3638 108
AJ001057.1 - 28 - 29 3581 108
AK092330.1 - 1 - 2 633 108
AK294195.1 - 2 - 3 252 108
AK299870.1 - 29 - 30 3791 108
BC114570.1 - 27 - 28 3634 108
BC115736.1 - 28 - 29 3670 108
BX538010.1 - 26 - 27 3664 108

View Full Product Details