Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRCAM
Entrez Gene ID
4897
Gene Name
neuronal cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.7: 108147626 - 108456442 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037132.2 NP_001032209.1 7 - 8 882 128
NM_001193582.1 NP_001180511.1 10 - 11 1262 128
NM_005010.4 NP_005001.3 9 - 10 1244 128
XM_005250373.2 XP_005250430.1 10 - 11 1262 128
XM_005250383.2 XP_005250440.1 10 - 11 1262 128
XM_005250385.2 XP_005250442.1 10 - 11 1262 128
XM_006716003.1 XP_006716066.1 10 - 11 1262 128
XM_006716007.1 XP_006716070.1 10 - 11 1262 128
XM_006716012.1 XP_006716075.1 10 - 11 1262 128
XM_006716014.1 XP_006716077.1 10 - 11 1262 128
XM_011516253.1 XP_011514555.1 10 - 11 1262 128
XM_011516255.1 XP_011514557.1 9 - 10 1017 128
XM_011516256.1 XP_011514558.1 10 - 11 1262 128
XM_011516257.1 XP_011514559.1 9 - 10 1244 128
XM_011516258.1 XP_011514560.1 8 - 9 999 128
XM_011516259.1 XP_011514561.1 10 - 11 1262 128
XM_011516261.1 XP_011514563.1 9 - 10 1244 128
XM_011516265.2 XP_011514567.1 10 - 11 1262 128
XM_011516266.2 XP_011514568.1 10 - 11 1262 128
XM_011516267.1 XP_011514569.1 10 - 11 1262 128
XM_011516268.2 XP_011514570.1 8 - 9 1077 128
XM_011516269.1 XP_011514571.1 10 - 11 1262 128
XM_011516270.1 XP_011514572.1 10 - 11 1262 128
XM_011516271.1 XP_011514573.1 9 - 10 1244 128
XM_017012236.1 XP_016867725.1 10 - 11 1262 128
XM_017012237.1 XP_016867726.1 10 - 11 1262 128
XM_017012243.1 XP_016867732.1 10 - 11 1262 128
XM_017012244.1 XP_016867733.1 10 - 11 1262 128
XM_017012245.1 XP_016867734.1 10 - 11 1262 128
XM_017012246.1 XP_016867735.1 10 - 11 1262 128
XM_017012248.1 XP_016867737.1 10 - 11 1262 128
XM_017012251.1 XP_016867740.1 10 - 11 1262 128
XM_017012252.1 XP_016867741.1 10 - 11 1262 128
XM_017012253.1 XP_016867742.1 7 - 8 931 128
XM_017012254.1 XP_016867743.1 10 - 11 1262 128
XM_017012256.1 XP_016867745.1 7 - 8 931 128
XM_017012258.1 XP_016867747.1 9 - 10 986 128
XM_017012259.1 XP_016867748.1 7 - 8 931 128
GenBank mRNA AJ001057.1 - 7 - 8 761 128
AK299870.1 - 10 - 11 1250 128
BC098401.1 - 9 - 10 1232 128
BX538010.1 - 9 - 10 1189 128

View Full Product Details