The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297592.1 | NP_001284521.1 | 11 - 12 | 1937 | 127 |
NM_001297594.1 | NP_001284523.1 | 11 - 12 | 1959 | 127 | |
NM_015187.4 | NP_056002.2 | 11 - 12 | 2154 | 127 | |
XM_011513819.2 | XP_011512121.2 | 11 - 12 | 2148 | 127 | |
XM_017007923.1 | XP_016863412.1 | 10 - 11 | 1792 | 127 | |
GenBank mRNA | AB018289.1 | - | 10 - 11 | 1652 | 127 |
AK289687.1 | - | 11 - 12 | 1902 | 127 | |
AK300219.1 | - | 2 - 3 | 252 | 127 | |
BC009945.2 | - | 4 - 5 | 669 | 127 | |
BC029422.1 | - | 1 - 2 | 79 | 127 | |
BC041046.1 | - | 2 - 3 | 306 | 127 | |
BC060867.1 | - | 11 - 12 | 1947 | 127 | |
CR749817.1 | - | 11 - 12 | 1937 | 127 |