The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297592.1 | NP_001284521.1 | 4 - 5 | 962 | 71 |
NM_001297594.1 | NP_001284523.1 | 4 - 5 | 984 | 71 | |
NM_015187.4 | NP_056002.2 | 4 - 5 | 1179 | 71 | |
XM_011513819.2 | XP_011512121.2 | 4 - 5 | 1173 | 71 | |
XM_017007923.1 | XP_016863412.1 | 3 - 4 | 817 | 71 | |
GenBank mRNA | AB018289.1 | - | 3 - 4 | 677 | 71 |
AK289687.1 | - | 4 - 5 | 927 | 71 | |
BC060867.1 | - | 4 - 5 | 972 | 71 | |
CR749817.1 | - | 4 - 5 | 962 | 71 |