The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005731.3 | NP_005722.1 | 7 - 8 | 751 | 62 |
NM_152862.2 | NP_690601.1 | 8 - 9 | 846 | 62 | |
XM_011510471.1 | XP_011508773.1 | 4 - 5 | 1028 | 62 | |
XM_017003113.1 | XP_016858602.1 | 7 - 8 | 720 | 62 | |
XM_017003114.1 | XP_016858603.1 | 8 - 9 | 3129 | 62 | |
XM_017003115.1 | XP_016858604.1 | 7 - 8 | 710 | 62 | |
GenBank mRNA | AF006085.1 | - | 7 - 8 | 761 | 62 |
AK092520.1 | - | 4 - 5 | 1808 | 62 | |
AK093625.1 | - | 4 - 5 | 1799 | 62 | |
AK290268.1 | - | 8 - 9 | 745 | 62 | |
BC000590.2 | - | 8 - 9 | 709 | 62 | |
BT006898.1 | - | 7 - 8 | 677 | 62 | |
U50523.1 | - | 8 - 9 | 761 | 62 |