The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005731.3 | NP_005722.1 | 4 - 5 | 344 | 79 |
NM_152862.2 | NP_690601.1 | 5 - 6 | 439 | 79 | |
XM_017003113.1 | XP_016858602.1 | 4 - 5 | 313 | 79 | |
GenBank mRNA | AF006085.1 | - | 4 - 5 | 354 | 79 |
AK092520.1 | - | 1 - 2 | 1401 | 79 | |
AK290268.1 | - | 5 - 6 | 338 | 79 | |
BC000590.2 | - | 5 - 6 | 302 | 79 | |
BG282181.1 | - | 5 - 6 | 438 | 79 | |
BG765898.1 | - | 5 - 6 | 382 | 79 | |
BT006898.1 | - | 4 - 5 | 270 | 79 | |
U50523.1 | - | 5 - 6 | 354 | 79 |