The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197259.1 | NP_001184188.1 | 13 - 14 | 1515 | 91 |
NM_012381.3 | NP_036513.2 | 14 - 15 | 1613 | 91 | |
XM_017010633.1 | XP_016866122.1 | 14 - 15 | 1559 | 91 | |
XM_017010634.1 | XP_016866123.1 | 14 - 15 | 1559 | 91 | |
XM_017010635.1 | XP_016866124.1 | 13 - 14 | 1445 | 91 | |
XM_017010636.1 | XP_016866125.1 | 15 - 16 | 1617 | 91 | |
XM_017010637.1 | XP_016866126.1 | 15 - 16 | 1616 | 91 | |
XM_017010638.1 | XP_016866127.1 | 15 - 16 | 1617 | 91 | |
XM_017010639.1 | XP_016866128.1 | 15 - 16 | 1616 | 91 | |
XM_017010640.1 | XP_016866129.1 | 14 - 15 | 1502 | 91 | |
GenBank mRNA | AF093535.1 | - | 14 - 15 | 1537 | 91 |
AF125507.1 | - | 14 - 15 | 1546 | 91 | |
AF135044.1 | - | 14 - 15 | 1511 | 91 | |
AK222795.1 | - | 14 - 15 | 1539 | 91 | |
AK303056.1 | - | 14 - 15 | 1539 | 91 | |
AK303182.1 | - | 13 - 14 | 1334 | 91 | |
AK303195.1 | - | 13 - 14 | 1439 | 91 | |
AK315911.1 | - | 14 - 15 | 1500 | 91 | |
BC035494.2 | - | 14 - 15 | 1528 | 91 | |
U50950.1 | - | 10 - 11 | 1179 | 91 |