The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197259.1 | NP_001184188.1 | 16 - 17 | 1839 | 81 |
NM_012381.3 | NP_036513.2 | 17 - 18 | 1937 | 81 | |
NM_181837.2 | NP_862820.1 | 17 - 18 | 1940 | 81 | |
XM_005248704.1 | XP_005248761.1 | 17 - 18 | 1886 | 81 | |
XM_005248705.1 | XP_005248762.1 | 18 - 19 | 1944 | 81 | |
XM_005248706.1 | XP_005248763.1 | 18 - 19 | 1943 | 81 | |
XM_011535651.1 | XP_011533953.1 | 16 - 17 | 1772 | 81 | |
XM_017010632.1 | XP_016866121.1 | 17 - 18 | 1886 | 81 | |
XM_017010633.1 | XP_016866122.1 | 17 - 18 | 1883 | 81 | |
XM_017010634.1 | XP_016866123.1 | 17 - 18 | 1883 | 81 | |
XM_017010635.1 | XP_016866124.1 | 16 - 17 | 1769 | 81 | |
XM_017010636.1 | XP_016866125.1 | 18 - 19 | 1941 | 81 | |
XM_017010637.1 | XP_016866126.1 | 18 - 19 | 1940 | 81 | |
XM_017010638.1 | XP_016866127.1 | 18 - 19 | 1941 | 81 | |
XM_017010639.1 | XP_016866128.1 | 18 - 19 | 1940 | 81 | |
XM_017010640.1 | XP_016866129.1 | 17 - 18 | 1826 | 81 | |
GenBank mRNA | AF093535.1 | - | 17 - 18 | 1861 | 81 |
AF125507.1 | - | 17 - 18 | 1870 | 81 | |
AF135044.1 | - | 17 - 18 | 1835 | 81 | |
AK222795.1 | - | 17 - 18 | 1863 | 81 | |
AK303182.1 | - | 16 - 17 | 1658 | 81 | |
AK303195.1 | - | 16 - 17 | 1763 | 81 | |
AK315911.1 | - | 17 - 18 | 1824 | 81 | |
AL080116.1 | - | 17 - 18 | 1879 | 81 | |
BC035494.2 | - | 17 - 18 | 1852 | 81 | |
BC047689.1 | - | 17 - 18 | 1856 | 81 | |
U50950.1 | - | 13 - 14 | 1503 | 81 |