Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORC3
Entrez Gene ID
23595
Gene Name
origin recognition complex subunit 3
Gene Aliases
LAT, LATHEO, ORC3L
Chromosome Location
Chr.6: 87590067 - 87677824 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197259.1 NP_001184188.1 5 - 6 589 120
NM_012381.3 NP_036513.2 6 - 7 687 120
NM_181837.2 NP_862820.1 6 - 7 687 120
XM_005248704.1 XP_005248761.1 6 - 7 633 120
XM_005248705.1 XP_005248762.1 7 - 8 691 120
XM_005248706.1 XP_005248763.1 7 - 8 690 120
XM_011535651.1 XP_011533953.1 6 - 7 633 120
XM_011535652.2 XP_011533954.1 6 - 7 633 120
XM_017010632.1 XP_016866121.1 6 - 7 633 120
XM_017010633.1 XP_016866122.1 6 - 7 633 120
XM_017010634.1 XP_016866123.1 6 - 7 633 120
XM_017010635.1 XP_016866124.1 6 - 7 633 120
XM_017010636.1 XP_016866125.1 7 - 8 691 120
XM_017010637.1 XP_016866126.1 7 - 8 690 120
XM_017010638.1 XP_016866127.1 7 - 8 691 120
XM_017010639.1 XP_016866128.1 7 - 8 690 120
XM_017010640.1 XP_016866129.1 7 - 8 690 120
GenBank mRNA AF093535.1 - 6 - 7 611 120
AF125507.1 - 6 - 7 620 120
AF135044.1 - 6 - 7 585 120
AK222795.1 - 6 - 7 613 120
AK302750.1 - 7 - 8 681 120
AK303056.1 - 6 - 7 613 120
AK303195.1 - 5 - 6 513 120
AK315911.1 - 6 - 7 574 120
AK316298.1 - 6 - 7 618 120
AL080116.1 - 6 - 7 626 120
BC035494.2 - 6 - 7 602 120
BC047689.1 - 6 - 7 603 120
CR456814.1 - 1 - 2 120 120
U50950.1 - 2 - 3 253 120

View Full Product Details