Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSCR3
Entrez Gene ID
10311
Gene Name
DSCR3 arrestin fold containing
Gene Aliases
DCRA, DSCRA
Chromosome Location
Chr.21: 37223425 - 37268124 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005260909.4 XP_005260966.1 3 - 4 1179 65
GenBank mRNA AK300024.1 - 3 - 4 445 65
AK307465.1 - 2 - 3 190 65
AK316460.1 - 3 - 4 361 65

View Full Product Details