The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000188.2 | NP_000179.2 | 7 - 8 | 973 | 85 |
NM_001322364.1 | NP_001309293.1 | 9 - 10 | 1066 | 85 | |
NM_001322365.1 | NP_001309294.1 | 12 - 13 | 1371 | 85 | |
NM_001322366.1 | NP_001309295.1 | 7 - 8 | 864 | 85 | |
NM_001322367.1 | NP_001309296.1 | 6 - 7 | 877 | 85 | |
NM_033496.2 | NP_277031.1 | 7 - 8 | 970 | 85 | |
NM_033497.2 | NP_277032.1 | 10 - 11 | 1188 | 85 | |
NM_033498.2 | NP_277033.1 | 10 - 11 | 1242 | 85 | |
NM_033500.2 | NP_277035.2 | 11 - 12 | 1335 | 85 | |
XM_005269737.1 | XP_005269794.1 | 7 - 8 | 1244 | 85 | |
XM_011539732.1 | XP_011538034.1 | 11 - 12 | 1242 | 85 | |
GenBank mRNA | AB209526.1 | - | 7 - 8 | 969 | 85 |
AK091267.1 | - | 7 - 8 | 1098 | 85 | |
AK128226.1 | - | 12 - 13 | 1366 | 85 | |
AK292012.1 | - | 7 - 8 | 973 | 85 | |
AK294428.1 | - | 7 - 8 | 970 | 85 | |
AK309904.1 | - | 6 - 7 | 799 | 85 | |
BC008730.2 | - | 7 - 8 | 928 | 85 | |
M75126.1 | - | 7 - 8 | 953 | 85 |