The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277115.1 | NP_001264044.1 | 53 - 54 | 8834 | 136 |
GenBank mRNA | AJ320497.1 | - | 53 - 54 | 8855 | 136 |