Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UACA
Entrez Gene ID
55075
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
NUCLING
Chromosome Location
Chr.15: 70654554 - 70778823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008224.1 NP_001008225.1 14 - 15 1493 131
NM_018003.2 NP_060473.2 14 - 15 1270 131
XM_005254529.4 XP_005254586.1 13 - 14 1487 131
XM_011521752.2 XP_011520054.2 14 - 15 1599 131
XM_011521753.2 XP_011520055.2 13 - 14 1567 131
XM_011521754.2 XP_011520056.1 13 - 14 1048 131
XM_017022394.1 XP_016877883.1 13 - 14 1319 131
XM_017022395.1 XP_016877884.1 14 - 15 1130 131
GenBank mRNA AB046781.2 - 14 - 15 1166 131
AF322916.1 - 14 - 15 1215 131
AK055435.1 - 14 - 15 1520 131
AL834243.1 - 13 - 14 1462 131
BC016881.1 - 14 - 15 1482 131
BC113407.1 - 14 - 15 1189 131
BC113409.1 - 14 - 15 1189 131
BC143266.1 - 14 - 15 1189 131
BC143267.1 - 11 - 12 862 131

View Full Product Details