The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008224.1 | NP_001008225.1 | 16 - 17 | 4287 | 142 |
NM_018003.2 | NP_060473.2 | 16 - 17 | 4064 | 142 | |
XM_005254529.4 | XP_005254586.1 | 15 - 16 | 4281 | 142 | |
XM_011521752.2 | XP_011520054.2 | 16 - 17 | 4393 | 142 | |
XM_011521753.2 | XP_011520055.2 | 15 - 16 | 4361 | 142 | |
XM_011521754.2 | XP_011520056.1 | 15 - 16 | 3842 | 142 | |
XM_017022394.1 | XP_016877883.1 | 15 - 16 | 4113 | 142 | |
XM_017022395.1 | XP_016877884.1 | 16 - 17 | 3924 | 142 | |
GenBank mRNA | AB046781.2 | - | 16 - 17 | 3960 | 142 |
AF322916.1 | - | 16 - 17 | 4009 | 142 | |
AL834464.1 | - | 1 - 2 | 421 | 142 | |
BC113407.1 | - | 16 - 17 | 3983 | 142 | |
BC113409.1 | - | 16 - 17 | 3983 | 142 | |
BC143266.1 | - | 16 - 17 | 3983 | 142 |