Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UACA
Entrez Gene ID
55075
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
NUCLING
Chromosome Location
Chr.15: 70654554 - 70778823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008224.1 NP_001008225.1 3 - 4 634 95
NM_018003.2 NP_060473.2 3 - 4 411 95
XM_005254529.4 XP_005254586.1 3 - 4 661 95
XM_011521752.2 XP_011520054.2 3 - 4 740 95
XM_011521753.2 XP_011520055.2 3 - 4 741 95
XM_011521754.2 XP_011520056.1 3 - 4 371 95
XM_017022394.1 XP_016877883.1 3 - 4 493 95
XM_017022395.1 XP_016877884.1 3 - 4 271 95
GenBank mRNA AB046781.2 - 3 - 4 307 95
AF322916.1 - 3 - 4 356 95
AK055435.1 - 3 - 4 661 95
AL834243.1 - 3 - 4 636 95
BC016881.1 - 3 - 4 623 95
BC113407.1 - 3 - 4 330 95
BC113409.1 - 3 - 4 330 95
BC143266.1 - 3 - 4 330 95
BC143267.1 - 3 - 4 330 95
CN303461.1 - 3 - 4 412 95

View Full Product Details