The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032595.4 | NP_115984.3 | 1 - 2 | 1378 | 73 |
XM_011525381.2 | XP_011523683.1 | 1 - 2 | 1888 | 73 | |
GenBank mRNA | AL713642.1 | - | 1 - 2 | 185 | 73 |