The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307923.1 | NP_001294852.1 | 7 - 8 | 964 | 114 |
NM_012397.3 | NP_036529.1 | 7 - 8 | 937 | 114 | |
XM_005266707.3 | XP_005266764.1 | 8 - 9 | 1241 | 114 | |
XM_011526029.2 | XP_011524331.1 | 7 - 8 | 888 | 114 | |
GenBank mRNA | AF169949.1 | - | 6 - 7 | 799 | 114 |
AJ001696.2 | - | 7 - 8 | 871 | 114 | |
AJ001697.2 | - | 7 - 8 | 839 | 114 | |
AJ001698.1 | - | 8 - 9 | 1018 | 114 | |
AK290323.1 | - | 7 - 8 | 911 | 114 | |
AK296797.1 | - | 7 - 8 | 921 | 114 | |
AK298049.1 | - | 5 - 6 | 616 | 114 | |
AK303867.1 | - | 5 - 6 | 648 | 114 | |
AK303979.1 | - | 7 - 8 | 828 | 114 | |
AK313486.1 | - | 7 - 8 | 909 | 114 | |
BC101821.1 | - | 7 - 8 | 826 | 114 | |
BC143402.1 | - | 7 - 8 | 853 | 114 |