Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC24A
Entrez Gene ID
10802
Gene Name
SEC24 homolog A, COPII coat complex component
Gene Aliases
-
Chromosome Location
Chr.5: 134648402 - 134727911 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252231.1 NP_001239160.1 1 - 2 387 84
NM_021982.2 NP_068817.1 1 - 2 387 84
XM_006714523.2 XP_006714586.1 1 - 2 762 84
XM_017008963.1 XP_016864452.1 1 - 2 762 84
GenBank mRNA AK304060.1 - 1 - 2 387 84
BC019341.1 - 1 - 2 348 84
CD109783.1 - 1 - 2 410 84

View Full Product Details