The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019104.2 | NP_061977.1 | 5 - 6 | 639 | 83 |
GenBank mRNA | AK293116.1 | - | 5 - 6 | 404 | 83 |
AK301992.1 | - | 5 - 6 | 639 | 83 | |
BC009071.1 | - | 5 - 6 | 358 | 83 | |
BI767008.1 | - | 5 - 6 | 389 | 83 |