The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006612.5 | NP_006603.2 | 10 - 11 | 1220 | 126 |
XM_005256424.2 | XP_005256481.1 | 11 - 12 | 1211 | 126 | |
GenBank mRNA | AB014606.2 | - | 10 - 11 | 1216 | 126 |
AK307401.1 | - | 10 - 11 | 1063 | 126 | |
BC034993.1 | - | 10 - 11 | 1194 | 126 | |
U91329.1 | - | 9 - 10 | 976 | 126 |