The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007344.3 | NP_031370.2 | 2 - 3 | 1437 | 68 |
NR_134525.1 | 2 - 3 | 1437 | 68 | ||
XM_006717273.3 | XP_006717336.2 | 2 - 3 | 1445 | 68 | |
XM_017015105.1 | XP_016870594.1 | 2 - 3 | 1944 | 68 | |
GenBank mRNA | AK302902.1 | - | 2 - 3 | 1423 | 68 |
BC143821.1 | - | 2 - 3 | 1410 | 68 |