Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTF1
Entrez Gene ID
7270
Gene Name
transcription termination factor 1
Gene Aliases
TTF-1, TTF-I
Chromosome Location
Chr.9: 132375548 - 132406851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205296.1 NP_001192225.1 3 - 4 476 124
NM_007344.3 NP_031370.2 4 - 5 1850 124
NR_134525.1 4 - 5 1850 124
XM_006717273.3 XP_006717336.2 4 - 5 1858 124
XM_017015105.1 XP_016870594.1 4 - 5 2357 124
GenBank mRNA AK302902.1 - 4 - 5 1836 124
BC127669.1 - 3 - 4 451 124
BC127670.1 - 3 - 4 451 124
BC143048.1 - 3 - 4 451 124
BC143049.1 - 3 - 4 451 124
BC143821.1 - 4 - 5 1823 124

View Full Product Details