The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016538.2 | NP_057622.1 | 9 - 10 | 1052 | 65 |
XM_006722284.2 | XP_006722347.1 | 7 - 8 | 786 | 65 | |
XM_011523580.1 | XP_011521882.1 | 8 - 9 | 918 | 65 | |
XM_011523581.2 | XP_011521883.1 | 8 - 9 | 2655 | 65 | |
GenBank mRNA | AF233395.1 | - | 9 - 10 | 1034 | 65 |
AK002027.1 | - | 7 - 8 | 1832 | 65 | |
AK131437.1 | - | 5 - 6 | 2235 | 65 | |
AK290265.1 | - | 9 - 10 | 1020 | 65 | |
AL137626.1 | - | 6 - 7 | 620 | 65 | |
BC017305.2 | - | 9 - 10 | 1016 | 65 | |
BC101791.1 | - | 9 - 10 | 1033 | 65 | |
BC101793.1 | - | 9 - 10 | 1032 | 65 |