The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015175.2 | NP_055990.1 | 19 - 20 | 3004 | 58 |
XM_006713072.3 | XP_006713135.1 | 18 - 19 | 2920 | 58 | |
XM_011533533.2 | XP_011531835.1 | 19 - 20 | 3001 | 58 | |
XM_017006010.1 | XP_016861499.1 | 19 - 20 | 3001 | 58 | |
XM_017006011.1 | XP_016861500.1 | 19 - 20 | 2857 | 58 | |
XM_017006012.1 | XP_016861501.1 | 18 - 19 | 2920 | 58 | |
XM_017006013.1 | XP_016861502.1 | 19 - 20 | 3001 | 58 | |
XM_017006014.1 | XP_016861503.1 | 18 - 19 | 2776 | 58 | |
XM_017006015.1 | XP_016861504.1 | 17 - 18 | 2632 | 58 | |
XM_017006016.1 | XP_016861505.1 | 14 - 15 | 3578 | 58 | |
GenBank mRNA | AB011112.3 | - | 7 - 8 | 1240 | 58 |
AK131104.1 | - | 19 - 20 | 2844 | 58 |