The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145396.1 | NP_001138868.1 | 14 - 15 | 2086 | 92 |
NM_153329.3 | NP_699160.2 | 15 - 16 | 2239 | 92 | |
XM_011526441.1 | XP_011524743.1 | 15 - 16 | 2057 | 92 | |
XM_011526442.1 | XP_011524744.1 | 15 - 16 | 2074 | 92 | |
GenBank mRNA | AK074136.1 | - | 14 - 15 | 2269 | 92 |
AK297101.1 | - | 14 - 15 | 2069 | 92 | |
AK298587.1 | - | 12 - 13 | 1821 | 92 | |
AY007096.1 | - | 8 - 9 | 1172 | 92 | |
BC014895.2 | - | 15 - 16 | 2199 | 92 | |
BC035641.1 | - | 2 - 3 | 366 | 92 | |
BC042142.1 | - | 8 - 9 | 1162 | 92 |