Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGSF3
Entrez Gene ID
3321
Gene Name
immunoglobulin superfamily member 3
Gene Aliases
EWI-3, LCDD, V8
Chromosome Location
Chr.1: 116574398 - 116667755 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001542.3 NP_001533.2 5 - 6 1911 112
XM_005270793.2 XP_005270850.1 6 - 7 2021 112
XM_006710593.3 XP_006710656.1 5 - 6 1322 112
XM_011541315.1 XP_011539617.1 6 - 7 2073 112
GenBank mRNA AF031174.1 - 5 - 6 1386 112

View Full Product Details