The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178002.2 | NP_001171473.1 | 7 - 8 | 777 | 65 |
NM_138813.3 | NP_620168.1 | 7 - 8 | 916 | 65 | |
XM_006722654.2 | XP_006722717.1 | 7 - 8 | 916 | 65 | |
XM_006722655.2 | XP_006722718.1 | 7 - 8 | 995 | 65 | |
XM_006722656.2 | XP_006722719.1 | 7 - 8 | 804 | 65 | |
XM_011527707.1 | XP_011526009.1 | 7 - 8 | 916 | 65 | |
XM_011527708.1 | XP_011526010.1 | 7 - 8 | 916 | 65 | |
XM_011527709.1 | XP_011526011.1 | 7 - 8 | 916 | 65 | |
XM_011527711.2 | XP_011526013.1 | 7 - 8 | 916 | 65 | |
GenBank mRNA | AK125225.1 | - | 7 - 8 | 698 | 65 |
AY302538.1 | - | 7 - 8 | 740 | 65 | |
BC035162.2 | - | 7 - 8 | 911 | 65 | |
BU542181.1 | - | 6 - 7 | 565 | 65 |