The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002054.4 | NP_002045.1 | 4 - 5 | 648 | 64 |
GenBank mRNA | BC005278.1 | - | 4 - 5 | 491 | 64 |
BT006813.1 | - | 3 - 4 | 392 | 64 | |
J04040.1 | - | 4 - 5 | 429 | 64 |